Learn More
BBD, MedChemExpress
MedChemExpress BBD(NSC240867; Benzylamino-NBD) is a biochemical reagent/chromogenic reagent.

745.27 DKK - 1994.10 DKK
Specifications
Chemical Name or Material | BBD |
---|---|
CAS | 18378-20-6 |
Purity Grade Notes | Research |
Molecular Formula | C13H10N4O3 |
Synonym | NSC240867 Benzylamino-NBD |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16496986
![]() |
Medchem Express
HY-15907 |
100 mg |
745.27 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16421796
![]() |
Medchem Express
HY-15907 |
500 mg |
1994.10 DKK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BBD(NSC240867Specifications
BBD | |
Research | |
NSC240867 Benzylamino-NBD | |
O=[N+](C1=CC=C(NCC2=CC=CC=C2)C3=NON=C31)[O-] | |
270.24 | |
Research | |
Solid |
18378-20-6 | |
C13H10N4O3 | |
DMSO : ≥ 34 mg/mL (125.81 mM) | |
270.24 | |
98.0% | |
Brown |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.