Learn More
Azlon™ Class B PMP Volumetric Flask
Manufactured from transparent PMP provides clear easy to read calibration line
Brand: Azlon™ FDF136N
This item is not returnable.
View return policy
Description
- Manufactured from transparent PMP offering excellent clarity
- Clear easy to read calibration line
- Calibrated to ISO 1042, class B tolerances
- Supplied with polypropylene stopper
Specifications
Transparent | |
PMP | |
1 | |
Autoclavable | |
Stopper |
500 mL | |
270 mm (with stopper) | |
±0.5 mL | |
B |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Azlon™ Class B PMP Volumetric Flask > Capacity: 500mL
Spot an opportunity for improvement?Share a Content Correction