Learn More
BRAND™ Dispenser, tilt measure without bottle
Tilt dispenser is best suited for dispensing suspensions.
Brand: BRAND™ 43070
546.14 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Dispenser head
- Calibrated to deliver (TD, Ex)
- Inscriptions in green enamel
- Particularly well-suited for dispensing suspensions
Specifications
Borosilicate Glass 3.3 | |
Without bottle | |
25 mL |
Accessories and Services
Optional Accessory
-
BRAND™ Joint Clip For Tilt Measure Dispenser Joint clip for dispensers, tilt measure ... Product Code: 10044040
68.60 DKK / Each -
BRAND™ Clear Reservoir Bottle Reservoir bottle, Boro 4.1, clear1000ml Product Code: 10145680
179.00 DKK / Each -
Brand™ Ground Joint Sleeves X10 Ground joint sleeve, PTFEfor NS 29/3... Product Code: 12055332
242.00 DKK / Pack of 10
Product Content Correction
Product Title
BRAND™ Dispenser, tilt measure without bottle > Capacity: 25 mL
Spot an opportunity for improvement?Share a Content Correction