missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atovaquone, Thermo Scientific™
560.00 DKK - 1495.00 DKK
Chemical Identifiers
CAS | 95233-18-4 |
---|---|
Molecular Formula | C22H19ClO3 |
Molecular Weight (g/mol) | 366.84 |
MDL Number | 00889188 |
InChI Key | BSJMWHQBCZFXBR-UHFFFAOYSA-N |
IUPAC Name | 3-[4-(4-chlorophenyl)cyclohexyl]-4-hydroxy-1,2-dihydronaphthalene-1,2-dione |
SMILES | OC1=C(C2CCC(CC2)C2=CC=C(Cl)C=C2)C(=O)C(=O)C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17575545
|
Thermo Scientific Acros
468762500 |
250 mg |
560.00 DKK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17565545
|
Thermo Scientific Acros
468760010 |
1 g |
1495.00 DKK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
95233-18-4 | |
366.84 | |
BSJMWHQBCZFXBR-UHFFFAOYSA-N | |
OC1=C(C2CCC(CC2)C2=CC=C(Cl)C=C2)C(=O)C(=O)C2=CC=CC=C12 |
C22H19ClO3 | |
00889188 | |
3-[4-(4-chlorophenyl)cyclohexyl]-4-hydroxy-1,2-dihydronaphthalene-1,2-dione |
Specifications
Glass Bottle | |
Available | |
C22H19ClO3 | |
Yellow to Dark Yellow | |
Soluble in THF; Slightly soluble in acetone; Insoluble in methanol and water | |
OC1=C(C2CCC(CC2)C2=CC=C(Cl)C=C2)C(=O)C(=O)C2=CC=CC=C12 | |
366.84 |
95233-18-4 | |
Powder | |
00889188 | |
221°C to 225°C | |
BSJMWHQBCZFXBR-UHFFFAOYSA-N | |
3-[4-(4-chlorophenyl)cyclohexyl]-4-hydroxy-1,2-dihydronaphthalene-1,2-dione | |
Atovaquone |
Safety and Handling
Recommended Storage : Normal conditions
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Atovaquone, Thermo Scientific™