missing translation for 'onlineSavingsMsg'
Få mere at vide

Antioxidant 24, TRC

Artikelnummer. 30138740
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138740 50 g
30136377 250 g
30011024 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30138740 Leverandør Toronto Research Chemicals Leverandørnr. TRCA69750550G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 26741-53-7
Molekylær formel C33 H50 O6 P2
Molekylvægt (g/mol) 604.69
Synonym Phosphorous acid, cyclic neopentanetetrayl bis(2,4-di-tert-butylphenyl) ester (8CI), 2,4,8,10-Tetraoxa-3,9-diphosphaspiro[5.5]undecane, 3,9-bis(2,4-di-tert-butylphenoxy)- (8CI), 3,9-Bis[2,4-bis(1,1-dimethylethyl)phenoxy]-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane, Phenol, 2,4-di-tert-butyl-, dihydrogen phosphite cyclic neopentanetetrayl ester (8CI), 3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane, 3,9-Bis(2,4-ditert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane, ADK Stab PEP 24, ADK Stab PEP 24G, AT 626, Alkanox P 24, Antioxidant 126, Antioxidant 24, Antioxidant 242, Antioxidant 3026, Antioxidant 626, Antioxidant 6260, Antioxidant 627A, Antioxidant TH 24, Bis(2,4-di-t-butylphenyl)pentaerythritol diphosphite, Bis(2,4-di-tert-butylphenyl) pentaerythritol diphosphite, Bis(2,4-di-tert-butylphenyl)pentraerythritol diphosphite, Bruggolen H 175, Cyclic neopentanetetrayl bis(2,4-di-tert-butylphenyl phosphite), Di(2,4-di-tert-butylphenyl)pentaerythritol diphosphite, Doverphos S 9432, H 175, Irgafos 126, Irgafos 126FF, Irgafos 626, Irganox 242, KY 626, Magnafloc 6260, Mark PEP 24, Mark PEP 24G, PEP 24, PEP 24G, PEP 24Q, PL 34, Pentaerythritol bis(2,4-di-tert-butylbenzyl) diphosphite, Pentaerythritol bis(2,4-di-tert-butylphenyl phosphite), Pentaerythritol bis(2,4-di-tert-butylphenyl) diphosphite, Phos 5, RC 626, Songnox 6260, Songnox 6260FF, Songnox 6260PW, Sonox 626, THP 24, UV 242, Ultranox 2879A, Ultranox 624, Ultranox 626, Ultranox 626A, Ultranox 626GE, Ultranox 627AV, Ultranox 640, Weston 624, Weston 626, Weston MDW 626, Yiphos 3026, Cyclic Neopentanetetrayl Bis(2,4-di-tert-butylphenyl) Ester Phosphorous Acid, 3,9-bis(2,4-Di-tert-butylphenoxy)- 2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane, 2,4-di-tert-Butylphenol Dihydrogen Phosphite Cyclic neopentanetetrayl Ester, ADK Stab PEP 24, ADK Stab PEP 24G, AT 626, Alkanox P 24, Antioxidant 126, Antioxidant 242, Antioxidant 626, Antioxidant 6260, Antioxidant TH 24, Bis(2,4-di-tert-butylphenyl) pentaerythritol diphosphite, Bis(2,4-di-tert-butylphenyl)pentraerythritol diphosphite, Cyclic neopentanetetrayl bis(2,4-di-tert-butylphenyl phosphite), Di(2,4-di-tert-butylphenyl)pentaerythritol diphosphite, Doverphos S 9432, Irgafos 126, Irgafos 126FF, Irganox 242, KY 626 Magnafloc 6260, Mark PEP 24, Mark PEP 24G, PEP 24, PEP 24G, PEP 24Q, PL 34 Pentaerythritol bis(2,4-di-tert-butylphenyl phosphite), Pentaerythritol bis(2,4-di-tert-butylphenyl) diphosphite, Phos 5, Songnox 6260, Songnox 6260PW, UV 242, Ultranox 2879A, Ultranox 624 Ultranox 626, Ultranox 626A, Ultranox 626GE, Ultranox 627AV, Ultranox 640, Weston 624, Weston 626, Weston MDW 626, Yiphos 3026, 3,9-Bis[2,4-bis(1,1-dimethylethyl)phenoxy]-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
IUPAC navn 3,9-bis(2,4-ditert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
SMIL CC(C)(C)c1ccc(OP2OCC3(COP(Oc4ccc(cc4C(C)(C)C)C(C)(C)C)OC3)CO2)c(c1)C(C)(C)C

Tekniske data

Kemisk navn eller materiale Antioxidant 24
Mængde 50 g
InChI formel InChI=1S/C33H50O6P2/c1-29(2,3)23-13-15-27(25(17-23)31(7,8)9)38-40-34-19-33(20-35-40)21-36-41(37-22-33)39-28-16-14-24(30(4,5)6)18-26(28)32(10,11)12/h13-18H,19-22H2,1-12H3
Formel vægt 604.3083
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.