Learn More
8 Component Mix in DMSO, SPEX CertiPrep™
390.00 DKK - 460.00 DKK
Specifications
Chemical Name or Material | 8 Component Mix in DMSO |
---|---|
Name Note | USP 467 Standard |
Packaging | Glass Ampoule |
CAS | 127-19-5, 68-12-2, 107-21-1, 872-50-4, 75-12-7 |
Explosion Limit | 1.8% (volume); 63% (volume) |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
16390498
|
SPEX Certiprep
USP-RS-C2B |
1 mL | Glass Ampoule |
390.00 DKK
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16310508
|
SPEX Certiprep
USP-RS-C2C |
1 mL | Glass Ampoule |
460.00 DKK
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
8 Component Mix in DMSO | |
Glass Ampoule | |
1.8% (volume); 63% (volume) | |
2.5 hPA (20°C) | |
N,N-Dimethylacetamide; N,N-Dimethylformamide; Ethylene glycol; 1-Methyl-2-pyrrolidinone; Formamide; more | |
Fluid | |
DMSO | |
USP |
USP 467 Standard | |
127-19-5, 68-12-2, 107-21-1, 872-50-4, 75-12-7 | |
270°C | |
USP 467: Residual Solvents/ Organic Volatile Impurities | |
1.1g/cm3 | |
N,N-Dimethylacetamide, N,N-Dimethylformamide, Ethylene Glycol, 1-Methyl-2-pyrrolidinone, Formamide | |
1 mL |
Safety and Handling
Recommended Storage : Freezer
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Stock VOA
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : dimethyl sulfoxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
8 Component Mix in DMSO, SPEX CertiPrep™