Learn More
8 Component Mix in DMSO, SPEX CertiPrep™
Brand: SPEX Certiprep USP-RS-C2C
View more versions of this product
Specifications
8 Component Mix in DMSO | |
Glass Ampoule | |
1.8% (volume); 63% (volume) | |
2.5 hPA (20°C) | |
N,N-Dimethylacetamide; N,N-Dimethylformamide; Ethylene glycol; 1-Methyl-2-pyrrolidinone; Formamide; more | |
Fluid | |
DMSO | |
USP |
USP 467 Standard | |
127-19-5, 68-12-2, 107-21-1, 872-50-4, 75-12-7 | |
270°C | |
USP 467: Residual Solvents/ Organic Volatile Impurities | |
1.1g/cm3 | |
N,N-Dimethylacetamide, N,N-Dimethylformamide, Ethylene Glycol, 1-Methyl-2-pyrrolidinone, Formamide | |
1 mL |
Safety and Handling
- Stock VOA
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : dimethyl sulfoxide,N,N-dimethylacetamide, N,N-dimethylformamide, N-methyl-2-pyrrolidone, formamide