Learn More
Hamilton™ 700 Series Microliter™ Carbon Analyzer Syringes
Syringes are designed for water analysis with Total Organic Carbon (TOC)
819.00 DKK - 988.00 DKK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | Borosilicate Glass (Barrel), Stainless Steel (Plunger) |
Diameter (English) Outer | 0.305 in. |
Diameter (Metric) Outer | 7.75 mm |
Max. Temperature (English) | 122°F |
Product Code | Brand | Volume (Metric) | Needle Gauge | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Product Type | Price | Quantity & Availability | |||
11582475
|
Hamilton™
80660 |
100 μL | 24 G | Microliter Syringe |
988.00 DKK
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11552355
|
Hamilton™
80550 |
50 μL | 24 G | Microliter Syringe |
820.00 DKK
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11502485
|
Hamilton™
80770 |
250 μL | 24 G | Microliter Syringe |
987.00 DKK
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11562355
|
Hamilton™
80551 |
50 μL | 26 G | Carbon Analyzer Syringe |
819.00 DKK
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Microliter syringes with a stainless-steel plunger that is hand-fitted to the glass barrel and designed for liquid sampling
- Syringes have a carbon analyzer (CA) termination
- Available in a 50 or 250μL volume
Specifications
Not Autoclavable | |
0.305 in. | |
122°F | |
50°F | |
10°C | |
Blunt |
Borosilicate Glass (Barrel), Stainless Steel (Plunger) | |
7.75 mm | |
50°C | |
3 | |
Ea. | |
Carbon Analyzer, Cemented |