Få mere at vide
5-Hydroxy Indapamide, TRC
Artikelnummer. 30133927
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133927 50 mg
30008554 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133927 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94345050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 126750-70-7
Molekylær formel C16 H16 Cl N3 O4 S
Molekylvægt (g/mol) 381.83
Synonym Benzamide, 3-(aminosulfonyl)-4-chloro-N-(2,3-dihydro-5-hydroxy-2-methyl-1H-indol-1-yl)-, 3-(Aminosulfonyl)-4-chloro-N-(2,3-dihydro-5-hydroxy-2-methyl-1H-indol-1-yl)benzamide, 5-Hydroxyindapamide
IUPAC navn 4-chloro-N-(5-hydroxy-2-methyl-2,3-dihydroindol-1-yl)-3-sulfamoylbenzamide
SMIL CC1Cc2cc(O)ccc2N1NC(=O)c3ccc(Cl)c(c3)S(=O)(=O)N
Tekniske data
Kemisk navn eller materiale 5-Hydroxy Indapamide
Mængde 50 mg
InChI formel InChI=1S/C16H16ClN3O4S/c1-9-6-11-7-12(21)3-5-14(11)20(9)19-16(22)10-2-4-13(17)15(8-10)25(18,23)24/h2-5,7-9,21H,6H2,1H3,(H,19,22)(H2,18,23,24)
Formel vægt 381.055