Få mere at vide
4-Chloro-4’-hydroxybenzophenone, TRC
Artikelnummer. 30136937
Skift visning
Mængde:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136937 50 g
30009814 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136937 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36670050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 42019-78-3
Molekylær formel C13 H9 Cl O2
Molekylvægt (g/mol) 232.66
Synonym (4-Chlorophenyl)(4-hydroxyphenyl)methanone, Fenofibrate Imp. A (EP)
IUPAC navn (4-chlorophenyl)-(4-hydroxyphenyl)methanone
SMIL Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2
Tekniske data
Kemisk navn eller materiale (4-Chlorophenyl)(4-hydroxyphenyl)methanone
Renhedsgrad noter HPLC
Mængde 50 g
InChI formel InChI=1S/C13H9ClO2/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8,15H
Formel vægt 232.0291
Procent renhed >95%