missing translation for 'onlineSavingsMsg'
Få mere at vide

4-(2-Fluorophenyl)tetrahydropyran-4-carbonitrile, 97+%, Thermo Scientific™

Artikelnummer. 15419775
undefined
Mængde:
250 mg
1 g
5 g
missing translation for 'unitSize'
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15419775

missing translation for 'mfr': Thermo Scientific Alfa Aesar H52380.03

Request Bulk or Custom Format

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 859164-45-7
Molekylær formel C12H12FNO
Molekylvægt (g/mol) 205.232
MDL nummer MFCD14690885
InChI nøgle JOYSNTZJNYCWMW-UHFFFAOYSA-N
Synonym 4-2-fluorophenyl tetrahydropyran-4-carbonitrile, 4-2-fluorophenyl tetrahydro-2h-pyran-4-carbonitrile, 4-2-fluorophenyl oxane-4-carbonitrile, 4-2-fluoro-phenyl-tetrahydro-pyran-4-carbonitrile, 2h-pyran-4-carbonitrile, 4-2-fluorophenyl tetrahydro
PubChem CID 49875771
IUPAC navn 4-(2-fluorophenyl)oxane-4-carbonitrile
SMIL C1COCCC1(C#N)C2=CC=CC=C2F

Specifications

Kemisk navn eller materiale 4-(2-Fluorophenyl)tetrahydropyran-4-carbonitrile
Mængde 1 g
Formel vægt 205.23
Procent renhed ≥97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.