Learn More
3M™ Versaflo™ M-400 Series PAPR Outer Shroud
Outer shrouds are replacement parts for use with M-400 helmets.
1135.00 DKK - 1795.00 DKK
Specifications
Assigned Protection Factor (APF) | 1,000 |
---|---|
Product Type | Outer Shroud |
For Use With (Application) | Cleaning, electrical, facility maintenance, painting, sanding, welding |
For Use With (Equipment) | M-400 helmet |
Product Code | Brand | Style | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Style | Price | Quantity & Availability | ||||||
15700078
|
3M™
7100137850 |
Flame Resistant |
1795.00 DKK
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
15790068
|
3M™
7100137905 |
High Durability |
1135.00 DKK
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
Description
- The M-446 Outer Shroud is a comfortable, low-linting outer shroud for 3M™ Versaflo M-400 Helmets
- Made from Nomex, the M-447 Outer Shroud is a flame resistant outer shroud for 3M™ Versaflo M-400 Helmets
- Made from Cordura Nylon, the M-448 Outer Shroud is a high durability outer shroud for 3M™ Versaflo M-400 Helmets
- 3M Catalog# M-445, M-446, M-447, M-448
Specifications
1,000 | |
Outer Shroud | |
Cleaning, electrical, facility maintenance, painting, sanding, welding | |
M-400 helmet |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
3M™ Versaflo™ M-400 Series PAPR Outer Shroud