Få mere at vide
3,5-Di-tert-butylphenol, TRC
Artikelnummer. 30134069
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134069 500 mg
30135360 50 mg
17963375 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134069 Leverandør Toronto Research Chemicals Leverandørnr. TRCD679065500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1138-52-9
Molekylær formel C14 H22 O
Molekylvægt (g/mol) 206.32
Synonym Phenol, 3,5-di-tert-butyl- (7CI,8CI), 3,5-Bis(1,1-dimethylethyl)phenol, 3,5-Bis(tert-butyl)phenol, 3,5-Di-t-butylphenol, 3,5-Di-tert-butylphenol, NSC 68209
IUPAC navn 3,5-ditert-butylphenol
SMIL CC(C)(C)c1cc(O)cc(c1)C(C)(C)C
Tekniske data
Kemisk navn eller materiale 3,5-Di-tert-butylphenol
Mængde 500 mg
InChI formel InChI=1S/C14H22O/c1-13(2,3)10-7-11(14(4,5)6)9-12(15)8-10/h7-9,15H,1-6H3
Formel vægt 206.1671