Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass 3-Way Stopcocks
369.00 DKK - 714.00 DKK
Product Code | Brand | Plug Bore | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Plug Bore | Includes | Price | Quantity & Availability | |||||
10777851
|
Lenz Laborglasinstrumente™
02501402 |
2.5 mm | Solid Glass plug and screw thread retaining device |
369.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
10088170
|
Lenz Laborglasinstrumente™
02542404 |
4 mm | PTFE plug and screw thread retaining device |
714.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
10509971
|
Lenz Laborglasinstrumente™
02541802 |
2.5 mm | PTFE plug and screw thread retaining device |
378.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
10612472
|
Lenz Laborglasinstrumente™
02501804 |
4 mm | Solid Glass plug and screw thread retaining device |
446.00 DKK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
10597911
|
Lenz Laborglasinstrumente™
02502404 |
4 mm | Solid Glass plug and screw thread Rubber retaining device |
531.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
Specifications
3-way Stopcock | |
Borosilicate Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass 3-Way Stopcocks