Få mere at vide
2-Phenylbutyric Acid-d5, TRC
Artikelnummer. 30424410
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424410 10 mg
30409343 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424410 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31960210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189708-92-6
Molekylær formel C10H7D5O2
Molekylvægt (g/mol) 169.23
Synonym 2-Phenylbutyric Acid D5
IUPAC navn 3,3,4,4,4-pentadeuterio-2-phenylbutanoic acid
SMIL [2H]C([2H])([2H])C([2H])([2H])C(C(=O)O)c1ccccc1
Tekniske data
Kemisk navn eller materiale 2-Phenylbutyric Acid-d5
Mængde 10 mg
InChI formel InChI=1S/C10H12O2/c1-2-9(10(11)12)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H,11,12)/i1D3,2D2
Formel vægt 169.1151