Få mere at vide
2-Methyl-3-phenylpropionaldehyde, TRC
Artikelnummer. 30476000
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476000 50 mg
30464939 100 mg
30475269 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476000 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22293350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5445-77-2
Molekylær formel C10H12O
Molekylvægt (g/mol) 148.2
Synonym 2-Methyl-3-phenylpropanal
IUPAC navn 2-methyl-3-phenylpropanal
SMIL CC(Cc1ccccc1)C=O
Tekniske data
Kemisk navn eller materiale 2-Methyl-3-phenylpropionaldehyde
Mængde 50 mg
InChI formel InChI=1S/C10H12O/c1-9(8-11)7-10-5-3-2-4-6-10/h2-6,8-9H,7H2,1H3
Formel vægt 148.0888