missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Methoxy-6-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Artikelnummer. 15485547
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
15485547 5 g 5g
15475547 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Product Code. 15485547 Supplier Thermo Scientific Alfa Aesar Supplier No. H32789.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119692-41-0
Molekylær formel C9H7F3O3
Molekylvægt (g/mol) 220.147
MDL nummer MFCD09832319
InChI nøgle IOJYMQXQDCJQBF-UHFFFAOYSA-N
Synonym 2-methoxy-6-trifluoromethyl benzoic acid, benzoic acid,2-methoxy-6-trifluoromethyl, benzoic acid, 2-methoxy-6-trifluoromethyl, acmc-1cez3, 6-methoxy-2 trifluoromethylbenzoic acid, 6-methoxy-2-trifluoromethylbenzoic acid, 6-methoxy-2-trifluoromethyl benzoic acid
PubChem CID 15017533
IUPAC navn 2-methoxy-6-(trifluoromethyl)benzoic acid
SMIL COC1=CC=CC(=C1C(=O)O)C(F)(F)F

Specifications

Kemisk navn eller materiale 2-Methoxy-6-(trifluoromethyl)benzoic acid
Smeltepunkt 131°C to 134°C
Mængde 5 g
Formel vægt 220.15
Procent renhed 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.