Få mere at vide
2,2'-Isopropylidenediphenol, TRC
Artikelnummer. 30009647
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009647 100 mg
30008850 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009647 Leverandør Toronto Research Chemicals Leverandørnr. TRCI822980100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7559-72-0
Molekylær formel C15H16O2
Molekylvægt (g/mol) 228.29
Synonym 2,2'-(1-methylethylidene)bisphenol, 2,2'-isopropylidenediphenol, 2,2-Bis(2-hydroxyphenyl)propane, 2,2'-Dian, o,o'-Dian, o-Diphenylolpropane;
IUPAC navn 2-[2-(2-hydroxyphenyl)propan-2-yl]phenol
SMIL OC1=CC=CC=C1C(C)(C)C2=C(O)C=CC=C2
Tekniske data
Kemisk navn eller materiale 2,2'-Isopropylidenediphenol
Mængde 100 mg
InChI formel InChI=1S/C15H16O2/c1-15(2,11-7-3-5-9-13(11)16)12-8-4-6-10-14(12)17/h3-10,16-17H,1-2H3
Formel vægt 228.115