Learn More
Labcon™ PerformR™ 15 mL Polypropylene Centrifuge Tubes
Centrifuge tubes are designed for everyday centrifugation and storage needs. Available with either a flat cap or a plug style cap.
233.00 DKK - 1410.00 DKK
Specifications
Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP |
---|---|
Closure Material | Polyethylene |
Closure Color | Purple |
Capacity (Metric) | 15 mL |
Disposable | Single-use |
Product Code | Brand | Closure Type | Sterility | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Type | Sterility | Quantity | Price | Quantity & Availability | ||||
16674042
![]() |
Labcon™
3146-335-016-9 |
Flat Screw Cap | Sterile | 25 Tubes/Reusable Rack, 2 Racks/Pk., 6 Pk./Cs. |
1355.00 DKK
Pack of 300 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||
16654032
![]() |
Labcon™
3136-345-006-9 |
Flat Screw Cap | Sterile | 50 Tubes/Bag, 10 Bags/Cs. |
1060.00 DKK
Pack of 500 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
16684042
![]() |
Labcon™
3146-870-006-9 |
Flat Screw Cap | Non-sterile | 500 Tubes/Bag, 500 Caps in Separate Bag, 500 Tubes and Caps/Cs. |
1345.00 DKK
Pack of 500 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
||||
16664032
![]() |
Labcon™
3137-345-006-9 |
Plug Style Screw Cap | Sterile | 50 Tubes/Bag, 10 Bags/Cs. |
1290.00 DKK
Pack of 500 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||
16694042
![]() |
Labcon™
3147-870-006-9 |
Plug Style Screw Cap | Non-sterile | 500 Tubes/Bag, 500 Caps in Separate Bag, 500 Tubes and Caps/Cs. |
1410.00 DKK
Pack of 500 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
17517906
|
Labcon™
3147-335-016-9 |
Plug Style Screw Cap | Sterile | 25 Tubes/Rack, 2 Racks/Pk. |
233.00 DKK
Pack of 50 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||
Description
- Conical centrifuge tube
- ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
- Made in USA
Specifications
ISO 9001:2015; FDA CFR21 GMP | |
Purple | |
Single-use | |
Centrifuges, Rotators, Shakers | |
Yes | |
Autoclavable | |
118 mm | |
Yes | |
Yes | |
Yes |
Polyethylene | |
15 mL | |
Polypropylene | |
PerformR™ | |
Yes | |
Cylindrical | |
ATP, RNase/DNase and Endotoxin-free | |
Conical | |
Yes | |
Centrifuge Tube |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Labcon™ PerformR™ 15 mL Polypropylene Centrifuge Tubes