Learn More
Keystone Cleanroom Products 1421B Tyvek™ Drawstring Closure Bag, 1 to 5 in. Wide
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. These filling needle bags stay in place via the adjustable Tyvek™ drawstring closure system.
16650.00 DKK - 44390.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Certificate of Conformance (COC) |
Color | White |
Disposable | Single-use |
For Use With (Application) | Steam Sterilization |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
30020182
|
Keystone Cleanroom Products
6STK0102TD |
1 x 2 in. |
19605.00 DKK
Pack of 500 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
30020022
|
Keystone Cleanroom Products
6STK02-503TD |
2.5 x 3 in. |
27375.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020192
|
Keystone Cleanroom Products
6STK0203TD |
3 x 2 in. |
24420.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020276
|
Keystone Cleanroom Products
6STK0403TD |
3 x 4 in. |
27745.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020228
|
Keystone Cleanroom Products
6STK0204TD |
4 x 2 in. |
27375.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020279
|
Keystone Cleanroom Products
6STK0404TD |
4 x 4 in. |
28115.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020167
|
Keystone Cleanroom Products
6STK0409TD |
4 x 9 in. |
31815.00 DKK
Pack of 500 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||||
30020216
|
Keystone Cleanroom Products
6STK0105TD |
5 x 1 in. |
20350.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020086
|
Keystone Cleanroom Products
6STK0205TD |
5 x 2 in. |
28115.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020087
|
Keystone Cleanroom Products
6STK0305TD |
5 x 3 in. |
26265.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020210
|
Keystone Cleanroom Products
6STK0505TD18 |
5 x 5 in. |
18130.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020368
|
Keystone Cleanroom Products
6STK0106TD |
6 x 1 in. |
20350.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020098
|
Keystone Cleanroom Products
6STK0206TD |
6 x 2 in. |
28855.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020181
|
Keystone Cleanroom Products
6STK0306TD |
6 x 3 in. |
28115.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020195
|
Keystone Cleanroom Products
6STK0506TD |
6 x 5 in. |
16650.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020311
|
Keystone Cleanroom Products
6STK0107TD |
7 x 1 in. |
20720.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020329
|
Keystone Cleanroom Products
6STK0207TD |
7 x 2 in. |
26265.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020035
|
Keystone Cleanroom Products
6STK0108TD |
8 x 1 in. |
21455.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020337
|
Keystone Cleanroom Products
6STK0208TD |
8 x 2 in. |
38470.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020237
|
Keystone Cleanroom Products
6STK0308TD |
8 x 3 in. |
32920.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020253
|
Keystone Cleanroom Products
6STK0508TD |
8 x 5 in. |
16835.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30019995
|
Keystone Cleanroom Products
6STK0209TD |
9 x 2 in. |
39580.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020234
|
Keystone Cleanroom Products
6STK0309TD |
9 x 3 in. |
33295.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020076
|
Keystone Cleanroom Products
6STK0509TD |
9 x 5 in. |
31630.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020173
|
Keystone Cleanroom Products
6STK0210TD |
10 x 2 in. |
40320.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020151
|
Keystone Cleanroom Products
6STK0310TD |
10 x 3 in. |
17015.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020215
|
Keystone Cleanroom Products
6STK0410TD |
10 x 4 in. |
31815.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020178
|
Keystone Cleanroom Products
6STK0510TD |
10 x 5 in. |
17570.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020088
|
Keystone Cleanroom Products
6STK0111TD |
11 x 1 in. |
22570.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020265
|
Keystone Cleanroom Products
6STK0211TD |
11 x 2 in. |
41060.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020268
|
Keystone Cleanroom Products
6STK0511TD |
11 x 5 in. |
17945.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020067
|
Keystone Cleanroom Products
6STK0212TD |
12 x 2 in. |
21455.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
30020003
|
Keystone Cleanroom Products
6STK0312TD |
12 x 3 in. |
34400.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
¿Ya tiene una cuenta? Inicie sesión para ver su precio y disponibilidad. ¿Es nuevo aquí? ¡Regístrese hoy mismo! |
|||||
30020042
|
Keystone Cleanroom Products
6STK0512TD |
12 x 5 in. |
18130.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
¿Ya tiene una cuenta? Inicie sesión para ver su precio y disponibilidad. ¿Es nuevo aquí? ¡Regístrese hoy mismo! |
|||||
30020238
|
Keystone Cleanroom Products
6STK0213TD |
13 x 2 in. |
44390.00 DKK
Pack of 500 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
¿Ya tiene una cuenta? Inicie sesión para ver su precio y disponibilidad. ¿Es nuevo aquí? ¡Regístrese hoy mismo! |
|||||
30020340
|
Keystone Cleanroom Products
6STK0314TD |
14 x 3 in. |
17200.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
¿Ya tiene una cuenta? Inicie sesión para ver su precio y disponibilidad. ¿Es nuevo aquí? ¡Regístrese hoy mismo! |
|||||
30020073
|
Keystone Cleanroom Products
6STK0515TD |
15 x 5 in. |
19235.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
¿Ya tiene una cuenta? Inicie sesión para ver su precio y disponibilidad. ¿Es nuevo aquí? ¡Regístrese hoy mismo! |
|||||
30020027
|
Keystone Cleanroom Products
6STK0418TD |
18 x 4 in. |
18685.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
¿Ya tiene una cuenta? Inicie sesión para ver su precio y disponibilidad. ¿Es nuevo aquí? ¡Regístrese hoy mismo! |
|||||
30020360
|
Keystone Cleanroom Products
6STK0425TD |
25 x 4 in. |
20720.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
¿Ya tiene una cuenta? Inicie sesión para ver su precio y disponibilidad. ¿Es nuevo aquí? ¡Regístrese hoy mismo! |
|||||
30020012
|
Keystone Cleanroom Products
6STK0426TD |
26 x 4 in. |
21085.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
¿Ya tiene una cuenta? Inicie sesión para ver su precio y disponibilidad. ¿Es nuevo aquí? ¡Regístrese hoy mismo! |
|||||
30020247
|
Keystone Cleanroom Products
6STK0428TD |
28 x 4 in. |
21455.00 DKK
Pack of 250 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
¿Ya tiene una cuenta? Inicie sesión para ver su precio y disponibilidad. ¿Es nuevo aquí? ¡Regístrese hoy mismo! |
|||||
Descripción
- Designed for steam sterilization
- Tyvek™ on both sides for maximum breathability during sterilization
- Eliminates need to cut material and the use of autoclave tape
- Fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Sealed seams eliminate sewing threads and greatly reduce particulates
- Resists tearing and punctures
- Superior microbial barrier
- Lint-free Category 1 (Helmke Drum)
- Manufactured and packaged in a ISO 6 cleanroom environment
- Certificates of Conformance sent with shipment
Especificaciones
Autoclavable | |
White | |
Steam Sterilization | |
Tyvek | |
Up to 250°F | |
Autoclave Bag |
Certificate of Conformance (COC) | |
Single-use | |
Pharmaceutical Equipment and Components | |
Non-sterile | |
Up to 125°C |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.