Få mere at vide
1-(4-Hydrazinylbenzyl)-1H-1,2,4-triazole, TRC
Artikelnummer. 30132395
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132395 25 mg
30138456 2.5 mg
30006315 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132395 Leverandør Toronto Research Chemicals Leverandørnr. TRCH53243025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 212248-62-9
Molekylær formel C9H11N5 . 2HCl
Molekylvægt (g/mol) 189.22
Synonym 1-[(4-Hydrazinylphenyl)methyl]-1H-1,2,4-triazole Hydrochloride (1:2)
IUPAC navn [4-(1,2,4-triazol-1-ylmethyl)phenyl]hydrazine;dihydrochloride
SMIL Cl.Cl.NNc1ccc(Cn2cncn2)cc1
Tekniske data
Kemisk navn eller materiale 1-(4-Hydrazinylbenzyl)-1H-1,2,4-triazole
Mængde 25 mg
InChI formel InChI=1S/C9H11N5.2ClH/c10-13-9-3-1-8(2-4-9)5-14-7-11-6-12-14;;/h1-4,6-7,13H,5,10H2;2*1H
Formel vægt 261.0548