CAS RN 918407-35-9
CAS RN 918407-35-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Avoralstat, MedChemExpress
MedChemExpress Avoralstat (BCX4161), a potent and orally active plasma kallikrein (PKK) inhibitor, is used for hereditary angioedema research.
