CAS RN 90-49-3
CAS RN 90-49-3
IUPAC Name:
(2-phenylbutanoyl)urea
Synonyms:
pheneturide ethylphenacemide (pheneturide (inn))
Molecular Weight (g/mol):
206.25
Molecular Formula:
C11H14N2O2
InChi Key:
AJOQSQHYDOFIOX-UHFFFAOYNA-N
SMILES:
CCC(C(=O)NC(N)=O)C1=CC=CC=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Pheneturide
CAS: 90-49-3 Molecular Formula: C11H14N2O2 Molecular Weight (g/mol): 206.25 InChI Key: AJOQSQHYDOFIOX-UHFFFAOYNA-N IUPAC Name: (2-phenylbutanoyl)urea SMILES: CCC(C(=O)NC(N)=O)C1=CC=CC=C1
2
Phenylethylacetylurea (2-Phenylbutyrylurea), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.