CAS RN 885278-07-9
CAS RN 885278-07-9
IUPAC Name:
2-ethoxypyridine-3-carbaldehyde
Synonyms:
2-ethoxypyridine-3-carbaldehyde
Molecular Weight (g/mol):
151.17
Molecular Formula:
C8H9NO2
InChi Key:
WHEFRNNNELKVRQ-UHFFFAOYSA-N
SMILES:
CCOC1=C(C=O)C=CC=N1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2-Ethoxypyridine-3-carboxaldehyde, 97%
CAS: 885278-07-9 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.17 MDL Number: MFCD06804559 InChI Key: WHEFRNNNELKVRQ-UHFFFAOYSA-N Synonym: 2-ethoxynicotinaldehyde,2-ethoxypyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 2-ethoxy,2-ethoxy-3-pyridinecarboxaldehyde,acmc-20ac3a,2-ethoxy-pyridine-3-carbaldehyde PubChem CID: 25067327 IUPAC Name: 2-ethoxypyridine-3-carbaldehyde SMILES: CCOC1=C(C=O)C=CC=N1