accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 880635-03-0

H3CONHOONCH3NHOFFFH3C(E/Z)

CAS RN 880635-03-0

IUPAC Name: N-(3-{4-[2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethoxy]phenyl}-2-({4-oxo-4-[4-(trifluoromethyl)phenyl]but-2-en-2-yl}amino)propyl)propanamide
Synonyms: N-(3-{4-[2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethoxy]phenyl}-2-({4-oxo-4-[4-(trifluoromethyl)phenyl]but-2-en-2-yl}amino)propyl)propanamide
Molecular Weight (g/mol): 619.69
Molecular Formula: C35H36F3N3O4
InChi Key: TYEFSRMOUXWTDN-UHFFFAOYNA-N
SMILES: CCC(=O)NCC(CC1=CC=C(OCCC2=C(C)OC(=N2)C2=CC=CC=C2)C=C1)NC(C)=CC(=O)C1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.