accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 7783-48-4

FFSr

CAS RN 7783-48-4

IUPAC Name: strontium(2+) difluoride
Synonyms: strontium fluoride
Molecular Weight (g/mol): 125.62
Molecular Formula: F2Sr
InChi Key: FVRNDBHWWSPNOM-UHFFFAOYSA-L
SMILES: [F-].[F-].[Sr++]
Physical Form 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
Percent Purity 
  • (3)

Narrow Results

Narrow Results

  • (10)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.