CAS RN 6723-30-4
CAS RN 6723-30-4
IUPAC Name:
O-(oxan-2-yl)hydroxylamine
Synonyms:
O-(oxan-2-yl)hydroxylamine
Molecular Weight (g/mol):
117.15
Molecular Formula:
C5H11NO2
InChi Key:
NLXXVSKHVGDQAT-UHFFFAOYNA-N
SMILES:
NOC1CCCCO1