accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 6399-81-1

PBrHPhPhPh

CAS RN 6399-81-1

IUPAC Name: hydrogen triphenylphosphane bromide
Synonyms: hydrogen triphenylphosphine bromide
Molecular Weight (g/mol): 343.2
Molecular Formula: C18H16BrP
InChi Key: CMSYDJVRTHCWFP-UHFFFAOYSA-N
SMILES: [H+].[Br-].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.