CAS RN 6318-51-0
CAS RN 6318-51-0
IUPAC Navn:
2-(4-chlorobenzoyl)pyridine
Synonymer:
2-(4-chlorobenzoyl)pyridine
Molekylvægt (g/mol):
217.65
Molekylformel:
C12H8ClNO
InChi Key:
KHXSJSBQIWAIEG-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C(=O)C1=CC=CC=N1
1
–
2
af
2
Resultater
1
2-(4-Chlorobenzoyl)pyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(4-Chlorobenzoyl)pyridine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.