CAS RN 60088-54-2
CAS RN 60088-54-2
IUPAC Navn:
(²H₄)benzene-1,4-dicarboxylic acid
Synonymer:
(²H₄)benzene-1,4-dicarboxylic acid
Molekylvægt (g/mol):
170.16
Molekylformel:
C8H6O4
InChi Key:
KKEYFWRCBNTPAC-RHQRLBAQSA-N
SMILES:
[2H]C1=C([2H])C(=C([2H])C([2H])=C1C(O)=O)C(O)=O
1
–
3
af
3
Resultater
1
Terephthalic-d4 Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Terephthalic-2,3,5,6-d4 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
Terephthalic-d4 Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.