Få mere at vide
Terephthalic-d4 Acid, TRC
Artikelnummer. 30413410
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30413410 5 g
30411756 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413410 Leverandør Toronto Research Chemicals Leverandørnr. TRCT1125045G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Terephthalic acid ring-d4
CAS 60088-54-2
Renhedsgrad noter HPLC
Molekylær formel C8 2H4 H2 O4
Mængde 5 g
Synonym 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid (9CI','ACI), Terephthalic-d4 acid (6CI), 2,3,5,6-Tetradeuterioterephthalic acid, Terephthalic-2,3,5,6-d4 acid, Terephthalic-2,3,5,6-D4 acid
InChI formel InChI=1S/C8H6O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h1-4H,(H,9,10)(H,11,12)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C(=O)O)c([2H])c([2H])c1C(=O)O
IUPAC navn 2,3,5,6-tetradeuterioterephthalic acid
Molekylvægt (g/mol) 170.16
Formel vægt 170.0517
Procent renhed >95