Få mere at vide
Terephthalic-2,3,5,6-d4 Acid, CDN
Artikelnummer. 30322203
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30322203 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322203 Leverandør CDN Leverandørnr. CDND01611G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Terephthalic acid ring-d4
CAS 60088-54-2
Molekylær formel C8 2H4 H2 O4
Mængde 1 g
InChI formel InChI=1 S/C8H6O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h1-4 H,(H,9,10)(H,11,12)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(C(=O)O)c([2 H])c([2 H])c1C(=O)O
IUPAC navn 2,3,5,6-tetradeuterioterephthalic acid
Molekylvægt (g/mol) 170.16
Formel vægt 170.0517 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity