accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 57292-44-1

CH3H3CCH3OONH(R)ClHOO

CAS RN 57292-44-1

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-chlorophenyl)propanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(4-chlorophenyl)propanoate
Molecular Weight (g/mol): 298.74
Molecular Formula: C14H17ClNO4
InChi Key: BETBOAZCLSJOBQ-LLVKDONJSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C([O-])=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.