accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 541-47-9

CH3CH3OHO

CAS RN 541-47-9

IUPAC Name: 3-methylbut-2-enoic acid
Synonyms: β,β-dimethacrylate senecioic acid kyselina 3-methyl-2-butenova senecic acid b,b-dimethylacrylic acid senecate b,b-dimethylacrylate senecioate
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: YYPNJNDODFVZLE-UHFFFAOYSA-N
SMILES: CC(C)=CC(O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.