CAS RN 53-19-0
CAS RN 53-19-0
IUPAC Navn:
1-chloro-4-[2,2-dichloro-1-(2-chlorophenyl)ethyl]benzene
Synonymer:
mitotane (o,p)-DDD mytotan bristol myers squibb brand of mitotane O,p'-DDD khlodithan mitotane (product) o,p-TDE o,p-DDD lysodren
Molekylvægt (g/mol):
320.03
Molekylformel:
C14H10Cl4
InChi Key:
JWBOIMRXGHLCPP-UHFFFAOYNA-N
SMILES:
ClC(Cl)C(C1=CC=C(Cl)C=C1)C1=CC=CC=C1Cl
1
–
7
af
7
Resultater
2
Mitotane
CAS: 53-19-0 Molekylær formel: C14H10Cl4 Molekylvægt (g/mol): 320.03 InChI nøgle: JWBOIMRXGHLCPP-UHFFFAOYNA-N IUPAC navn: 1-chlor-4-[2,2-dichlor-1-(2-chlorphenyl)ethyl]benzen SMIL: ClC(Cl)C(C1=CC=C(Cl)C=C1)C1=CC=CC=C1Cl
3
Mitotane, MedChemExpress
MedChemExpress Mitotane(2,4′-DDD), an isomer of DDD and derivative of DDT, is an antineoplastic medication used in the treatment of adrenocortical carcinoma.
