CAS RN 504433-23-2
CAS RN 504433-23-2
IUPAC Navn:
3-[(1-methyl-1H-indol-3-yl)methylidene]-1H,2H,3H-pyrrolo[3,2-b]pyridin-2-one
Synonymer:
3-[(1-methylindol-3-yl)methylidene]-1H-pyrrolo[3,2-b]pyridin-2-one
Molekylvægt (g/mol):
275.31
Molekylformel:
C17H13N3O
InChi Key:
NXNQLECPAXXYTR-UHFFFAOYSA-N
SMILES:
CN1C=C(C=C2C(=O)NC3=C2N=CC=C3)C2=CC=CC=C12
1
–
2
af
2
Resultater
1
Tocris Bioscience™ GW 441756
CAS: 504433-23-2 Molekylær formel: C17H13N3O
2
GW 441756, MedChemExpress
MedChemExpress GW 441756 is a potent and specific nerve growth factor (NGF) receptor tyrosine kinases A (TrkA) inhibitor (IC50=2 nM), which eliminates the BmK NSPK-induced neurite outgrowth.