CAS RN 481-42-5
CAS RN 481-42-5
IUPAC Name:
5-hydroxy-2-methyl-1,4-dihydronaphthalene-1,4-dione
Synonyms:
plumbagin plumbagine plumbagone plumbaein
Molecular Weight (g/mol):
188.18
Molecular Formula:
C11H8O3
InChi Key:
VCMMXZQDRFWYSE-UHFFFAOYSA-N
SMILES:
CC1=CC(=O)C2=C(O)C=CC=C2C1=O
1
–
2
of
2
results
2
Medchem Express Plumbagin, MedChemExpress
MedChemExpress Plumbagin (2-Methyljuglone) is a naphthoquinone isolated from Plumbago zeylanica L, exhibits anticancer and antiproliferative activities.
