accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 453-72-5

CH3SOOFNOO

CAS RN 453-72-5

IUPAC Name: 1-fluoro-4-methanesulfonyl-2-nitrobenzene
Synonyms: 1-fluoro-4-methanesulfonyl-2-nitrobenzene
Molecular Weight (g/mol): 219.19
Molecular Formula: C7H6FNO4S
InChi Key: OUSNDSFSTBZESM-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.