accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 26027-38-3

H3COOH

CAS RN 26027-38-3

IUPAC Name: 2-(4-nonylphenoxy)ethan-1-ol
Synonyms: nonoxynol amway all purpose spray adjuvant akyporox NP 150 polyoxyethylene (18) nonyl phenyl ether liponox ncy polyoxyethylene nonylphenyl ether nonoxinols patentex ovals ovals, patentex nonylphenoxypolyethoxyethanols
Molecular Weight (g/mol): 264.41
Molecular Formula: C17H28O2
InChi Key: KUXGUCNZFCVULO-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CC=C(OCCO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.