CAS RN 2588-04-7
IUPAC Navn:
O,O-diethyl {[(ethanesulfonyl)methyl]sulfanyl}phosphonothioate
Synonymer:
phorate sulfone
Molekylvægt (g/mol):
292.36
Molekylformel:
C7H17O4PS3
InChi Key:
YVPSNUIHHFTTRL-UHFFFAOYSA-N
SMILES:
CCOP(=S)(OCC)SCS(=O)(=O)CC
1
–
3
af
3
Resultater
1
Phoratsulfon, TRC
CAS: 2588-04-7 Molekylær formel: C7 H17 O4 P S3 Molekylvægt (g/mol): 292.38 Synonym: Phosphorodithioic acid, O,O-diethyl (ethylsulfonyl)methyl ester (6CI),Methanethiol, (ethylsulfonyl)-, S-ester with O,O-diethyl phosphorodithioate (8CI),O,O-Diethyl S-ethylsulfonylmethyl phosphorodithioate,O,O-Diethyl S-ethylsulfonylmethyl thionophosphate,Phorate sulfone,Phosphorodithioic acid, O,O-diethyl S-[(ethylsulfonyl)methyl] ester,Phorate Sulfone IUPAC navn: diethoxy-(ethylsulfonylmethylsulfanyl)-sulfanyliden-λ^{5}-fosfan SMIL: CCOP(=S)(OCC)SCS(=O)(=O)CC