CAS RN 25014-41-9
CAS RN 25014-41-9
IUPAC Navn:
prop-2-enenitrile
Synonymer:
acrylonitrile caswell no. 010 miller fumigrain acritet ccris 8 epa pesticide chemical code 000601 VCN nitrile acrilico cyanure de vinyle akrylonitryl
Molekylvægt (g/mol):
53.06
Molekylformel:
C3H3N
InChi Key:
NLHHRLWOUZZQLW-UHFFFAOYSA-N
SMILES:
C=CC#N
1
–
4
af
4
Resultater
2
Polyacrylonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Polyacrylonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Polyacrylonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.