CAS RN 24851-98-7
CAS RN 24851-98-7
IUPAC Navn:
methyl 2-(3-oxo-2-pentylcyclopentyl)acetate
Synonymer:
methyl dihydrojasmonate sodium tetrahydrojasmonate
Molekylvægt (g/mol):
226.32
Molekylformel:
C13H22O3
InChi Key:
KVWWIYGFBYDJQC-UHFFFAOYNA-N
SMILES:
CCCCCC1C(CC(=O)OC)CCC1=O
1
–
3
af
3
Resultater
1
Methyl (3-Oxo-2-pentylcyclopentyl)acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl (3-Oxo-2-pentylcyclopentyl)acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl (3-Oxo-2-pentylcyclopentyl)acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.