CAS RN 2461-42-9
CAS RN 2461-42-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
alpha-Naphthyl Glycidyl Ether, 90%, TRC
CAS: 2461-42-9 Molekylær formel: C13 H12 O2 Molekylvægt (g/mol): 200.23 Synonym: 1-(1-Naphthyloxy)-2,3-epoxypropane IUPAC navn: 2-(naphthalen-1-yloxymethyl)oxirane SMIL: C(Oc1cccc2ccccc12)C3CO3
2
1-(1-Naphthyloxy)-2,3-epoxypropane, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.