CAS RN 2459-09-8
CAS RN 2459-09-8
IUPAC Navn:
methyl pyridine-4-carboxylate
Synonymer:
I-nicotinic acid, methyl ester
Molekylvægt (g/mol):
137.14
Molekylformel:
C7H7NO2
InChi Key:
OLXYLDUSSBULGU-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=NC=C1
1
–
3
af
3
Resultater
1
Methyl Isonicotinate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl Isonicotinate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl Isonicotinate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.