CAS RN 24579-73-5
CAS RN 24579-73-5
IUPAC Navn:
propyl N-[3-(dimethylazaniumyl)propyl]carbamate chloride
Synonymer:
propamocarb hcl
Molekylvægt (g/mol):
224.73
Molekylformel:
C9H21ClN2O2
InChi Key:
MKIMSXGUTQTKJU-UHFFFAOYSA-N
SMILES:
[Cl-].CCCOC(=O)NCCC[NH+](C)C
1
–
5
af
5
Resultater
2
Propamocarb, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Propamocarb, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Propamocarb, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Propamocarb 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality