accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 23943-96-6

CH3O(R)CH3OO

CAS RN 23943-96-6

IUPAC Name: (2R)-2-methoxypropanoate
Synonyms: (2R)-2-methoxypropanoate
Molecular Weight (g/mol): 103.1
Molecular Formula: C4H7O3
InChi Key: ICPWFHKNYYRBSZ-GSVOUGTGSA-M
SMILES: CO[C@H](C)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.