accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 214262-86-9

OHFCl

CAS RN 214262-86-9

IUPAC Name: 2-(2-chloro-6-fluorophenyl)ethan-1-ol
Synonyms: 2-(2-chloro-6-fluorophenyl)ethanol
Molecular Weight (g/mol): 174.6
Molecular Formula: C8H8ClFO
InChi Key: UYZAUXBNFFFZFA-UHFFFAOYSA-N
SMILES: OCCC1=C(F)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.