accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 186692-46-6

CH3(R)HONHNHNNNCH3H3CN

CAS RN 186692-46-6

IUPAC Name: (2R)-2-{[6-(benzylamino)-9-(propan-2-yl)-9H-purin-2-yl]amino}butan-1-ol
Synonyms: seliciclib
Molecular Weight (g/mol): 354.46
Molecular Formula: C19H26N6O
InChi Key: BTIHMVBBUGXLCJ-OAHLLOKOSA-N
SMILES: CC[C@H](CO)NC1=NC(NCC2=CC=CC=C2)=C2N=CN(C(C)C)C2=N1
Color 
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.