missing translation for 'onlineSavingsMsg'
Learn More

Roscovitine
GREENER_CHOICE

Potent, selective cyclin-dependent kinase inhibitor

1975.00 DKK

Specifications

CAS 186692-46-6
Molecular Formula C19H26N6O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18767506
GREENER_CHOICE
missing translation for 'viewDocuments'
Tocris Bioscience
1332/10
10 mg
1975.00 DKK
10mg
Please to purchase this item. Need a web account? Register with us today!
18777136
GREENER_CHOICE
missing translation for 'viewDocuments'
Tocris Bioscience
1332/50
50mg N/A
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclin-dependent kinase (cdk) inhibitor. Displays sub-micromolar inhibition of cdk1, cdk2, cdk5, cdk7 and cdk9 (reported IC50 values range from 0.16 - 0.28 ÎĽM for cdk5 to 0.8 ÎĽM for cdk7). Selective for cdks over a wide range of related kinases including ERK1 and ERK2. Arrests L1210 cells in G1 phase. Inhibits phosphorylation of vimentin in vivo. Antimitotic.
Specifications

Specifications

186692-46-6
C19H26N6O
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Roscovitine

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.