accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1617-90-9

H3C(S)NN(S)(S)OHOOCH3

CAS RN 1617-90-9

IUPAC Name: methyl (15S,17S,19S)-15-ethyl-17-hydroxy-1,11-diazapentacyclo[9.6.2.0²,⁷.0⁸,¹⁸.0¹⁵,¹⁹]nonadeca-2,4,6,8(18)-tetraene-17-carboxylate
Synonyms: minorin vincachron arteriovinca vinodrel retard vinkametrin vincamidol pervincamine vincaminum vincamine vincamina
Molecular Weight (g/mol): 354.45
Molecular Formula: C21H26N2O3
InChi Key: RXPRRQLKFXBCSJ-GIVPXCGWSA-N
SMILES: CC[C@@]12CCCN3CCC4=C([C@H]13)N(C1=CC=CC=C41)[C@](O)(C2)C(=O)OC
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.